9-Xanthencarbonsäure

Lieferant: Thermo Scientific

Synonyme: Xanthen-9-carbonsäure

MAYBRH00001.10EA 0 EUR
MAYBRH00001.10 MAYBRH00001.50 MAYBRH00001.1
9-Xanthencarbonsäure
9-Xanthencarbonsäure
Formel: C₁₄H₁₀O₃
Molekulargewicht: 226,23 g/mol
MDL Number: MFCD00005059
CAS-Nummer: 82-07-5
EINECS: 201-394-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR