Indol-3-carbonitril
Lieferant: Molekula
Synonyme:
3-Indolcarbonitril
, 3-Cyanindol
, 1H-Indol-3-carbonitril
, 3-Cyan-1H-indol
MOLEM37090532EA
0
EUR
MOLEM37090532
Indol-3-carbonitril
Indol-3-carbonitril
Formel:
C₉H₆N₂ Molekulargewicht: 142,16 g/mol Siedepunkt: 350 °C (1013 hPa) |
MDL Number:
MFCD00022717 CAS-Nummer: 5457-28-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...