7-Indolcarbonsäure

Lieferant: Molekula

MOLEM53645945EA 0 EUR
MOLEM53645945
7-Indolcarbonsäure
7-Indolcarbonsäure
Formel: C₉H₇NO₂
Molekulargewicht: 161,16 g/mol
MDL Number: MFCD00210442
CAS-Nummer: 1670-83-3
EINECS: 216-801-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR