3-Hydroxyindol

Lieferant: Molekula

MOLE46347275EA 0 EUR
MOLE46347275
3-Hydroxyindol
3-Hydroxyindol
Formel: C₈H₇NO
Molekulargewicht: 133,15 g/mol
CAS-Nummer: 480-93-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR