9-Methylxanthin

Lieferant: Molekula

Synonyme: 3,7-Dihydro-9-methyl-1H-purin-2,6-dion9-Methyl-1H,3H-purin-2,6-dion9-Methyl-3,7-dihydro-1H-purin-2,6-dion9-Methyl-2,6(1H,3H)-purindion9-Methyl-3,7-dihydro-2H-purin-2,6-dion2,6-Dihydroxy-9-methylpurin

MOLEM32091016EA 0 EUR
MOLEM32091016
9-Methylxanthin
9-Methylxanthin
Formel: C₆H₆N₄O₂
Molekulargewicht: 166,14 g/mol
CAS-Nummer: 1198-33-0
EINECS: 214-831-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR