Digital tensiometer, TD 4
Lieferant: Lauda
LAUDLMT860EA 0 EUR
LAUDLMT860
Digital tensiometer, TD 4
Tensiometer
Digital tensiometer for surface and interfacial tension.
  • Precise and reproducible for research and quality testing
  • User-friendly with predefined methods
  • Precise high-resolution distance measurements
  • Intuitive graphic user interface and accurate temperature control
The ring/plate method of liquids is an established method for research, quality controls and products development in the beverage industry.
Weighing resolution: 0,1 mg
Zertifizierungen: ASTM D971, EN 14210, EN 14370, IEC 62961, EN 14059.