Kanamycinsulfat, ultrarein

Lieferant: Thermo Scientific
Gefahr

J17924.14EA 207 EUR
J17924.14 J17924.06
Kanamycinsulfat, ultrarein
Kanamycin sulphate

A selectable marker for cosmid vectors used in transfection of mammalian cells.


It binds to ribosomal components and inhibits protein synthesis. Kanamycin is inactivated by an aminoglycoside phospho transferase (MW 25,000) in the periplasmic space.

Formel: C₁₈H₃₈N₄O₁₅S
Molekulargewicht: 582,6 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD00070253
CAS-Nummer: 25389-94-0
EINECS: 246-933-9

Bestellen

Spezifikation Testergebnisse

Identity (A, B, C) Passes Test
Potency ≥750 µg of C₁₈H₃₆N₄O₁₁/mg powder
Residue on Ignition ≤1.0%
Impurity Content Organic Impurities Meets the Requirements
Loss on Drying ≤4.0%
pH (1%, H₂O) 6.5 - 8.5

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR