Precision quick response thermometer, type K, VWR® TD 111
620-1931EA 169 EUR
VWRI620-1931
Precision quick response thermometer, type K, VWR® TD 111
Precision quick response thermometer. Quick response measurement on surfaces, in liquids, air/gases...
  • LCD display approx. 13 mm high (31/2 digit)
  • Measuring interval: approx. 3 meas./sec
  • Battery life: approx. 2000 operating hours
Lieferumfang: Supplied with 9 volt battery and manual.