Edge tested lanyard, DBI-SALA® EZ-Stop™
Lieferant: 3M
CPSD1245555EA 159.25 EUR
CPSD1245555
Edge tested lanyard, DBI-SALA® EZ-Stop™
The fall arrest solution for all users working at height exposed to falls around or over edges.
  • Leg material 12 mm diameter kernmantle rope
  • Rope of 12 mm diameter offers extreme durability whilst maintaining flexible handling for ease of use
  • Orange accenting indicates suitability for edge applications
Body connecter made from aluminium triple lock (22 mm opening size, 25 kN breaking strength) or aluminium (21 mm opening size, 22 kN breaking strength).
Anchor connector made from aluminium (60 mm opening, 22 kN breaking strength) or aluminium triple Lock (22 mm opening size, 25 kN breaking strength).
Zertifizierungen: EN 355