Vertical lifeline system, Cabloc® Pro, 3M™
CPSD6191019EA 0 EUR
CPSD6191019 CPSD6191038 CPSD6191002
Vertical lifeline system, Cabloc® Pro, 3M™
The Cabloc Pro Vertical Lifeline System includes a 8 mm cable which is terminated at the top by a cable ferrule and cable clamps at the bottom. The cable is supported at 10 m intervals with intermediate cable guides. Quick and easy to install, to any ladder and/or pylon pole. The Cabloc traveller is lightweight and incorporates a shock absorber.
  • Includes an integrated energy absorber
  • Does not require a system energy absorber to be installed at the top mounting bracket
  • Allows for hands free travel over intermediate bypass cable guides
  • Anti-inversion feature - prevents user from fitting the traveller incorrectly
Weight capacity: 150 kg