Tetramethylrhodamine-5-isothiocyanate (5-TRITC), Sigma-Aldrich®
Lieferant: SIGMA-ALDRICH MICROSCOPY
M-Clarity™ quality level = MQ200
T0820-5MGEA
366
EUR
T0820-5MG
Tetramethylrhodamine-5-isothiocyanate (5-TRITC), Sigma-Aldrich®
Tetramethylrhodamine-5-isothiocyanate (5-TRITC)
Formel:
C₂₅H₂₁N₃O₃S Molekulargewicht: 443,52 g/mol Lagerungstemperatur: Gefrierschrank |
MDL Number:
MFCD07438035 CAS-Nummer: 80724-19-2 |
Spezifikation Testergebnisse
Appearance (Color) | Pink to Purple |
Appearance (Form) | Powder |
Proton NMR spectrum | Conforms to Structure |
EMM (in MeOH) | 82 - 98 (540 - 546 nm) |
Fluorescence (Emission max) | 567 - 575 nm |
Purity (HPLC) | ≥ 85 % at 254 nm |
Purity (HPLC) | ≥ 97% at 254 nm (Isomer purity) |