Meine Sitzung geöffnet lassen?
Endet in 
Ihre Sitzung ist abgelaufen
Möchten Sie sich wieder anmelden?
Ultra-low temperature freezers, UF V 700
Lieferant: Binder
BIND9020-0348EA 14980 EUR InStock
BIND9020-0348 BIND9020-0350
Ultra-low temperature freezers, UF V 700
Gefriergeräte Ultratiefkühlgeräte
These ultra low temperature freezers ensure the safe storage of samples at –80 °C. These combine outstanding environmental friendliness with low energy consumption, convenient operation and an individual safety concept.
  • Range of temperature controller –86 to –40 °C
  • Lowest energy consumption in its class
  • Powerful cascade compressor cooling unit
  • Environmentally friendly refrigerants R-290 and R-170
  • Efficient thermal insulation with vacuum insulation panels
  • Zero-voltage alarm contact for in-house alarm system
  • Features Ethernet interface and elapsed time indicator
The inner chamber, shelves, and inner doors are made entirely from stainless steel and are rust-proof. The door can be opened manually using an ergonomic door latch handle or at the touch of a button using an electromechanical door locking mechanism. Electronic access control is available via the NumPad option. Two rear access ports of Ø 28 mm are available.
The innovative door gasket concept reduces the tendency for ice to build up on the door frame and, at the same time, makes it easier to remove ice.
Bestellen
BESTELLEN
TECHNISCHE DATEN
ZUBEHÖR
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge