COLORCATCH 3 is the colorimeter which measures and transmits via Bluetooth the true colour directly to an iPod touch, an iPhone or an iPad. It allows to instantly apply the true colour directly to a picture (building, wall, dress, object) taken by the iPhone.
- Simple to use, only one button
- Standard delivered with more than 2100 colours
- Suitable for measuring on rough surfaces
- Independent of colour or paint manufacturers
- Capacity of 2000 measurements
- With 9 V alkaline type 6LR61 battery
Ideal for protective coatings, corrosion control, coating laboratories, paint production, surface finishing, powder coating, decorative coatings, building maintenance.
Languages: English, German, French, Italian, Spanish, Portuguese, Dutch, Turkish, Czech, Slovenian, Catalonian, Finnish and Polish.
Lieferumfang: Includes COLORCATCH 3 with RAL and NCS colours, nylon belt holder, battery, manual, three calibration plates and USB cable.