Methane in helium, 100 ppm, analytical standards Supelco®
Lieferant: Merck
SUPL501468EA 358 EUR
SUPL501468 SUPL307200
Methane in helium, 100 ppm, analytical standards Supelco®
A 40 μl injection of the Supelco® Methane Standard will provide more accurate flow measurements than the use of smaller quantities of a more concentrated methane sample.