Organic reference standard, Formaldehyde-2,4-DNPH, analytical standard, Supelco®
Lieferant: Merck
SUPL442597EA 132 EUR
SUPL442597
Organic reference standard, Formaldehyde-2,4-DNPH, analytical standard, Supelco®
Formaldehyde 2,4-dinitro­phenyl­hydrazone is a planar molecule and the nitro groups are almost coplanar with the benzene ring.