Organic reference standards, Menaquinone (Vitamin K2), analytical standard, Supelco®
Lieferant: Merck
47774.EA 35.5 EUR
47774.
Organic reference standards, Menaquinone (Vitamin K2), analytical standard, Supelco®
Standards Organische Standards
Menaquinone (K2), a vitamin K2 homologue with four isoprene units. It has been reported to enhance bone formation and plays a vital role in the production of blood coagulation factors.
Menaquinone may be used as a reference standard for the determination of menaquinone in human plasma using liquid chromatography with tandem mass spectrometry.