Nitrogen generator, Genius 1050

Lieferant: PEAK SCIENTIFIC
PEAK10-9554EA 0 EUR
PEAK10-9554
Nitrogen generator, Genius 1050
Gasgeneratoren Stickstoff-Generatoren
Utilizing Carbon Molecular Sieve and Pressure Swing Adsorption technology, the Genius 1050 provides on-site nitrogen for most LC-MS instruments. The Genius 1050 is well suited for a range of particularly demanding applications where specific performance specifications are required.

  • Compressor based solution, no need for an external air supply
  • Highly economical source of nitrogen gas with low lifetime running costs
  • The latest generation of compressors located in an insulated chamber reducing noise and vibration
  • Gas is supplied on demand so generator works to your schedule
  • 1 year comprehensive on-site warranty
  • Frequency: 50/60 Hz

Producing a consistent and dependable source of laboratory-grade nitrogen for LC-MS, the Genius 1050 is a high performance generator flexible enough to meet the specifi cations of a wide variety of LC-MS instruments.

Current: 7 A

Outlet pressure: 6,8 bar

Zertifizierungen: Conforms with: Low Voltage Directive 2014/35/EU
EN 61010-1: 2010 Electrical Equipment for measurement, control and laboratory use.
Electromagnetic Compatibility Directive 2014/30/EU
EN 61326-1: 2013 Electrical Equipment for measurement, control and laboratory use.
FCC47 CFR Part 15 Class B
Unintentional radiators; Conducted and Radiated emissions limits.
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR