Cerium Standard for AAS, TraceCERT®, Supelco®
Lieferant: Merck
TraceCERT®
53378-250MLEA 35.7 EUR
53378-250ML
Cerium Standard for AAS, TraceCERT®, Supelco®
Standards Kationischer Standards Cer-Standards
Zertifizierungen: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034 (ISO Guide 34)