Lithium standard for ICP-MS, TraceCERT®, Supelco®
Lieferant: Merck
SUPL39185-100MLEA 89.7 EUR
SUPL39185-100ML
Lithium standard for ICP-MS, TraceCERT®, Supelco®
Zertifizierungen: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034 (ISO Guide 34)