Titanium Standard for ICP-MS, TraceCERT®, Supelco®
Lieferant: Merck
TraceCERT®
44266-100MLEA 155 EUR
44266-100ML
Titanium Standard for ICP-MS, TraceCERT®, Supelco®
Standards Kationischer Standards Titan-Standards
Zertifizierungen: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034 (ISO Guide 34)