Calcium saccharin, Hyrdrat ≥98,0% (durch Trockensubstanz) FCC
Lieferant: Spectrum Chemical
SPCMC1146-12KGEA
6140
EUR
SPCMC1146-12KG
SPCMC1146-2.5KG
SPCMC1146-125GM
SPCMC1146-500GM
Calcium saccharin, Hyrdrat ≥98,0% (durch Trockensubstanz) FCC
Calcium saccharin
Calcium Saccharin, FCC is an artificial sweetener that is about 300 times as sweet as table sugar (sucrose).
Packaged in laboratory size bottles to production drum quantities and manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formel:
C₁₄H₈CaN₂O₆S₂·3½H₂O Lagerungstemperatur: Raumtemperatur |
CAS-Nummer:
6381-91-5 |
Spezifikation Testergebnisse
Assay (Anhydrous Basis) | 98.0 - 101.0 % |
Benzoate and Salicylate | To Pass Test |
Lead | max. 2 mg/kg |
Readily Carbonizable Substances | To Pass Test |
Selenium | max. 0.003 % |
Toluenesulphonamides | To Pass Test |
Water | Max. 15% |