Dimethyl-5-aminoisophthalat
Lieferant: Molekula
Synonyme:
5-Aminoisophthalsäuredimethylester
, 5-Amino-1,3-benzoldicarbonsäuredimethylester
MOLEM86296751EA
0
EUR
MOLEM86296751
Dimethyl-5-aminoisophthalat
Dimethyl-5-aminoisophthalat
Formel:
C₁₀H₁₁NO₄ Molekulargewicht: 209,2 g/mol |
MDL Number:
MFCD00008435 CAS-Nummer: 99-27-4 EINECS: 202-743-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...