Dodecyltrimethylammoniumbromid

Lieferant: Molekula

MOLEM14637765EA 0 EUR
MOLEM14637765
Dodecyltrimethylammoniumbromid
Dodecyltrimethylammonium bromide
Formel: C₁₅H₃₄BrN
Molekulargewicht: 308,35 g/mol
Schmelzpunkt: 264…266 °C
MDL Number: MFCD00011767
CAS-Nummer: 1119-94-4
EINECS: 214-290-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR