Guardian 5000 magnetic stirrer
Lieferant: Ohaus
OHAU30500543EA 433 EUR
OHAU30500543
Guardian 5000 magnetic stirrer
Reliable magnetic stirrer with exceptional safety features for everyday use.
  • Powerful, consistent stirring performance
  • Easy to read LCD display
  • Designed for durability and reliability
OHAUS Guardian 5000™ magnetic stirrers are designed for safety and simplicity in all research, academic and industrial applications.
The design ensures proper mixing and will ensure any spilt liquid is channeled away from the internal components. Ideal for everyday heating and stirring applications.