Safety spectacles, PW346
Lieferant: Portwest
Portwest®
PRTWPW34AMREA 1160 EUR
PRTWPW34AMR
Safety spectacles, PW346
Schutzbrillen
Frameless glasses, one piece curved lens with molded nose, molded browguard protection.
  • 99% UV protection
  • Anti-scratch coating for added durability
  • Individually packed for vending machines
  • Excellent side protection
Zertifizierungen: ANSI/ISEA Z87.1; EN 166 (1 F)
Lieferumfang: Cord included.