Delta™ harness with belt, DBI-SALA®, 3M™
Lieferant: 3M
DBI-SALA®
CPSD1112909EA 286.7 EUR
CPSD1112909 CPSD1112906 CPSD1112907 CPSD1112911 CPSD1112910
Delta™ harness with belt, DBI-SALA®, 3M™
Fallschutz Auffanggurte
The 3M DBI-SALA® Delta™ Harness with Belt provides an ergonomic fit and feel, with lightweight buckles and high strength webbing. It features a no-tangle triangular pattern, which creates a frame that allows the harness to be easily donned.
  • Water-repellent webbing helps keep users dry and comfortable
  • No-tangle design helps to give the correct fit of the harness
  • Abrasion-resistant webbing for lasting strength
  • Revolver vertical torso adjuster provides quick and easy adjustment
  • Stand up dorsal D-ring
  • Impact indicators make it simple to check the harness for damage and help ensure it’s safe to use
  • Covered identification labels
  • Universal sizing
Stay safe and secure when working at height with the DBI-SALA® Delta™ Harness with Belt. This safety harness features water-repellent webbing to keep the harness dry and to reduce the attraction of mould and dirt. The abrasion-resistant webbing has lasting strength and withstands severe demands, day in and day out. Revolver vertical torso adjusters provide simple and fast adjustments that eliminate loose ends and lock into place, preventing slippage.
The no-tangle design helps to give the correct fit of the harness while still providing additional comfort for everyday use. Impact indicators make it simple to check the harness for damage and help ensure it’s safe to use. Covered identification labels help protect important information for the life of the harness.