LR accelerator, Sigma-Aldrich®
Lieferant: Merck
62660-10ML-FEA 33.2 EUR
62660-10ML-F
LR accelerator, Sigma-Aldrich®
Färbemittel Mikroskopie- und Färbereagenzien
LR Accelerator is used for the chemical curing of LR White and LR Gold embedding media for routine and low temperature electron microscopy or the immunocytochemical localization of antigens by either light or electron microscopy.
  • Suitable for hematology
  • Suitable for histology