Calcimycin (mixed Mg/CA salt), Sigma-Aldrich®

Lieferant: SIGMA-ALDRICH MICROSCOPY
Achtung

C5149-5MGEA 182 EUR
C5149-5MG
Calcimycin (mixed Mg/CA salt), Sigma-Aldrich®
Calcimycin (mixed Mg/CA salt)

Ionophore highly selective for Ca2+. Potentiates responses to NMDA but not to quisqualate. In cell culture, stimulates nitric oxide production by calmodulin-dependent constitutive nitric oxide synthase.

Formel: (C₂₉H₃₆N₃O₆)₂·Mg(C₂₉H₃₆N₃O₆)₂·Ca
Lagerungstemperatur: Kühlschrank
CAS-Nummer: 52665-69-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR