JC-1, Sigma-Aldrich®

Lieferant: SIGMA-ALDRICH MICROSCOPY

T4069-5MGEA 733 EUR
T4069-5MG
JC-1, Sigma-Aldrich®
JC-1

A dual-emission potential-sensitive probe that can be used to measure mitochondrial membrane potential. JC-1 is a green-fluorescent (λex 520 nm) monomer at low membrane potential. At higher potentials, JC-1 forms red-fluorescent (λem 596 nm ) "J-aggregates," which exhibit broad excitation and very narrow emission spectra. The ratio of red to green fluorescence of JC-1 is dependent only on membrane potential, and not influenced by mitochondrial size, shape, or density.

Formel: C₂₅H₂₇Cl₄IN₄
Molekulargewicht: 652,223 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00188475
CAS-Nummer: 3520-43-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR