Dihydrofluorescein diacetate ≥97%, Sigma-Aldrich®
Lieferant: SIGMA-ALDRICH MICROSCOPY
292648-500MGEA 387 EUR
292648-500MG
Dihydrofluorescein diacetate ≥97%, Sigma-Aldrich®
Dihydrofluorescein diacetate
Dihydrofluorescein diacetate,97% (Diacetyldihydrofluorescein)
Formel: C₂₄H₁₈O₇
Schmelzpunkt: 213-215 °C (Lit.)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00005056
CAS-Nummer: 35340-49-9