Methylenblau-Lösung, Sigma-Aldrich®

Synonyme: Methylthioniniumchlorid3,7-Bis(dimethylamino)phenothiaziniumchloridN,N,N′,N′-Tetramethylthioninchlorid

319112-100MLEA 26.9 EUR
319112-100ML
Methylenblau-Lösung, Sigma-Aldrich®
Methylenblau

Methylene Blue solution has been used to study structural alterations and p53 immune-expression in rats.

Formel: C₁₆H₁₈ClN₃S
Molekulargewicht: 319,86 g/mol
Dichte: 0,98 g/mL (25 °C)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00150008
CAS-Nummer: 61-73-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR