Chemical protective tape, Tychem® 2000
Lieferant: DuPont
DUPOQC0990YLNLEA 694 EUR
DUPOQC0990YLNL
Chemical protective tape, Tychem® 2000
Chemical barrier tape with Tychem® 2000 level of protection and secure adhesion.
  • Chemical protection against inorganic acids and bases
  • Good flexibility for a smoother seal
  • Used in petrochemical applications, emergency response, chemical spill clean-up, maintenance operations
Permeation data available for 95 chemicals, allowing the comparison and match between the protection level of the chemical protective garment and the chemical glove.
Can be used with Type 3, 4, 5, 6 garments in less demanding chemical exposures to hold protective clothing in place and provide tighter seal at the connection points of PPE.
Zertifizierungen: Category I certified according to Regulation (EU) 2016/425.