(R)-Fmoc-β2-homophenylalanine

Lieferant: Apollo Scientific

APOSBICR140-500MGEA 1020 EUR
APOSBICR140-500MG APOSBICR140-1G
(R)-Fmoc-β2-homophenylalanine
(R)-Fmoc-β2-homophenylalanine
Formel: C₂₅H₂₃NO₄
Molekulargewicht: 401,454 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD07372498
CAS-Nummer: 828254-16-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR