1,2-Dichloro-4-benzenesulphonamido-5-nitrobenzene

Lieferant: Apollo Scientific

APOSBID2095-10GEA 30.6 EUR
APOSBID2095-10G APOSBID2095-50G
1,2-Dichloro-4-benzenesulphonamido-5-nitrobenzene
1,2-Dichloro-4-benzenesulphonamido-5-nitrobenzene
Formel: C₁₂H₈Cl₂N₂O₄S
Molekulargewicht: 347,174 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00127866
CAS-Nummer: 1596-36-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR