Amino-PEG5-CH2CO2H
|
Achtung
|
APOSBIPG1065-250MGEA
0
EUR
APOSBIPG1065-250MG
Amino-PEG5-CH2CO2H
Amino-PEG5-CH2CO2H
Formel:
C₁₂H₂₅NO₇ Molekulargewicht: 295,329 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD27977526 CAS-Nummer: 1263046-77-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...