Amino-PEG10-t-butyl ester
Lieferant: Apollo Scientific
APOSBIPG1078-250MGEA
671
EUR
APOSBIPG1078-250MG
Amino-PEG10-t-butyl ester
Amino-PEG10-t-butyl ester
Formel:
C₂₇H₅₅NO₁₂ Molekulargewicht: 585,725 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD26127812 CAS-Nummer: 1818294-42-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...