Bis-Mal-Lysine-PEG4-TFP ester
APOSBIPG1240-100MGEA
0
EUR
APOSBIPG1240-100MG
Bis-Mal-Lysine-PEG4-TFP ester
Bis-Mal-Lysine-PEG4-TFP ester
Formel:
C₃₇H₄₅F₄N₅O₁₃ Molekulargewicht: 843,773 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD28385461 CAS-Nummer: 2173083-46-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...