Bis-propargyl-PEG5
Lieferant: Apollo Scientific
|
Achtung
|
APOSBIPG1300-250MGEA
379
EUR
APOSBIPG1300-250MG
Bis-propargyl-PEG5
Bis-propargyl-PEG5
Formel:
C₁₄H₂₂O₅ Molekulargewicht: 270,321 g/mol Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD22201533 CAS-Nummer: 159428-42-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...