3-(2-Methoxyethoxy)propanoic acid

Synonyme: m-PEG2-acid

APOSBIPG1541-100MGEA 0 EUR
APOSBIPG1541-100MG
3-(2-Methoxyethoxy)propanoic acid
3-(2-Methoxyethoxy)propanoic acid
Formel: C₆H₁₂O₄
Molekulargewicht: 148,157 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD09939037
CAS-Nummer: 149577-05-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR