m-PEG7-aldehyde

Lieferant: Apollo Scientific

APOSBIPG1576-100MGEA 567 EUR
APOSBIPG1576-100MG
m-PEG7-aldehyde
m-PEG7-aldehyde
Formel: C₁₆H₃₂O₈
Molekulargewicht: 352,42 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD28976707
CAS-Nummer: 1058691-77-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR