m-PEG12-aldehyde

APOSBIPG1578-100MGEA 671 EUR
APOSBIPG1578-100MG
m-PEG12-aldehyde
m-PEG12-aldehyde
Formel: C₂₆H₅₂O₁₃
Molekulargewicht: 572,683 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD13185015
CAS-Nummer: 2185872-20-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR