exo-BCN-PEG4-acid

Achtung

APOSBIPG1887-100MGEA 0 EUR
APOSBIPG1887-100MG
exo-BCN-PEG4-acid
exo-BCN-PEG4-acid
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD29059947
CAS-Nummer: 2226472-38-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR