Z-1-Bromononadeca-4,7,10,13-tetraene

APOSBISC1007-2GEA 0 EUR
APOSBISC1007-2G APOSBISC1007-1G
Z-1-Bromononadeca-4,7,10,13-tetraene
Z-1-Bromononadeca-4,7,10,13-tetraene
Formel: C₁₉H₃₁Br
Molekulargewicht: 339,353 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00673587
CAS-Nummer: 117567-53-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR