Scandium(III) sulphate hydrate 99,99%

Achtung

APOSIN3154-5GEA 0 EUR
APOSIN3154-5G APOSIN3154-1G
Scandium(III) sulphate hydrate 99,99%
Scandium(III) sulphate hydrate
Formel: Sc₂(SO₄)₃·xH₂O
Molekulargewicht: 378,10 g/mol (anhy)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00149853
CAS-Nummer: 52788-54-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR