2-(4-Nitrophenyl)-2,3,4,5-tetrahydro-1H-1,5-benzodiazepine

APOSOR01592-50MGEA 0 EUR
APOSOR01592-50MG
2-(4-Nitrophenyl)-2,3,4,5-tetrahydro-1H-1,5-benzodiazepine
2-(4-Nitrophenyl)-2,3,4,5-tetrahydro-1H-1,5-benzodiazepine
Formel: C₁₅H₁₅N₃O₂
Molekulargewicht: 269,299 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD06245405
CAS-Nummer: 904813-76-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR