1-(2-Nitrobenzyl)piperazine dihydrochloride 97%

Lieferant: Apollo Scientific
Achtung

APOSOR0225-5GEA 241 EUR
APOSOR0225-5G
1-(2-Nitrobenzyl)piperazine dihydrochloride 97%
1-(2-Nitrobenzyl)piperazine dihydrochloride
Formel: C₁₁H₁₅N₃O₂·2ClH
Molekulargewicht: 294,178 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD04972579
CAS-Nummer: 827614-54-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR