2-(Aminomethyl)-3-chlorothiophene
Lieferant: Apollo Scientific
|
Gefahr
|
APOSOR103654-1GEA
423
EUR
APOSOR103654-1G
APOSOR103654-500MG
2-(Aminomethyl)-3-chlorothiophene
2-(Aminomethyl)-3-chlorothiophene
Formel:
C₅H₆ClNS Molekulargewicht: 147,626 g/mol Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD06409002 CAS-Nummer: 214759-25-8 UN-Nummer: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...