2-[(Phthalimid-1-yl)methyl]benzeneboronic acid, pinacol ester
Lieferant: Apollo Scientific
APOSOR10579-1GEA
263
EUR
APOSOR10579-1G
2-[(Phthalimid-1-yl)methyl]benzeneboronic acid, pinacol ester
2-[(Phthalimid-1-yl)methyl]benzeneboronic acid, pinacol ester
Formel:
C₂₁H₂₂BNO₄ Molekulargewicht: 363,215 g/mol Schmelzpunkt: 113…115 °C Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD02179492 CAS-Nummer: 380430-66-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...