N-Methoxy-N-methyl-2-(triphenylphosphoranylidene)acetamide

Lieferant: Apollo Scientific

APOSOR10665-5GEA 205 EUR
APOSOR10665-5G APOSOR10665-25G
N-Methoxy-N-methyl-2-(triphenylphosphoranylidene)acetamide
N-Methoxy-N-methyl-2-(triphenylphosphoranylidene)acetamide
Formel: C₂₂H₂₂NO₂P
Molekulargewicht: 363,389 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00134433
CAS-Nummer: 129986-67-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR