3-(3,3-Dimethylureido)phenylacetic acid
|
Achtung
|
APOSOR10865-1GEA
0
EUR
APOSOR10865-1G
3-(3,3-Dimethylureido)phenylacetic acid
3-(3,3-Dimethylureido)phenylacetic acid
Formel:
C₁₁H₁₄N₂O₃ Molekulargewicht: 222,24 g/mol Schmelzpunkt: 155…156 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD03411198 CAS-Nummer: 959577-84-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...